Learn More
Medchemexpress LLC Tamsulosin (hydrochloride) | 106463-17-6 | 99.8% | 1 ML
SDP
Catalog No. 5000408553
Encompass_Preferred
Tamsulosin hydrochloride is an orally active antagonist of the α1-adrenergic receptor. It is used in the research of prostatic hyperplasia and has been shown to induce apoptosis and attenuate abdominal aortic aneurysm growth and inflammation in animal models.
  • Exhibits intrinsic activity at human alpha1B adrenergic receptor (EC50 of 0.13 nM).
  • Attenuates high glucose-induced injury in human glomerular endothelial cells.
  • Influences ECM production and distribution, and cellular metabolism levels in ARPE 19 cells.
  • Induces apoptosis and clusterin expression in NRP-152 cells.
Catalog No. 50-004-08553 Supplier Medchemexpress LLC Supplier No. HYB0661A10MM/1ML
Tamsulosin hydrochloride is an orally active antagonist of the α1-adrenergic receptor. It is used in the research of prostatic hyperplasia and has been shown to induce apoptosis and attenuate abdominal aortic aneurysm growth and inflammation in animal models.
  • Exhibits intrinsic activity at human alpha1B adrenergic receptor (EC50 of 0.13 nM).
  • Attenuates high glucose-induced injury in human glomerular endothelial cells.
  • Influences ECM production and distribution, and cellular metabolism levels in ARPE 19 cells.
  • Induces apoptosis and clusterin expression in NRP-152 cells.