Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tamsulosin hydrochloride | 106463-17-6 | 99.8% | 100 MG
SDP

Catalog No. 5000407739
Encompass_Preferred

Tamsulosin hydrochloride is an orally active antagonist of the α1-adrenergic receptor. It induces apoptosis and is used for the research of prostatic hyperplasia. Tamsulosin hydrochloride also attenuates abdominal aortic aneurysm growth and inhibits inflammation in animal models.

  • Orally active α1-adrenergic receptor antagonist
  • Induces apoptosis
  • Used for prostatic hyperplasia research
  • Attenuates abdominal aortic aneurysm growth
  • Inhibits inflammation in animal models
  • Molecular weight: 444.97
  • Chemical formula: C20H29ClN2O5S
  • Appearance: Solid
  • Color: White to off-white
  • Recommended storage: 4°C, sealed, away from moisture and light
  • Solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)
  • Solubility in DMSO: 50 mg/mL (112.37 mM)

Catalog No. 50-004-07739 Supplier Medchemexpress LLC Supplier No. HYB0661A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tamsulosin hydrochloride is an orally active antagonist of the α1-adrenergic receptor. It induces apoptosis and is used for the research of prostatic hyperplasia. Tamsulosin hydrochloride also attenuates abdominal aortic aneurysm growth and inhibits inflammation in animal models.

  • Orally active α1-adrenergic receptor antagonist
  • Induces apoptosis
  • Used for prostatic hyperplasia research
  • Attenuates abdominal aortic aneurysm growth
  • Inhibits inflammation in animal models
  • Molecular weight: 444.97
  • Chemical formula: C20H29ClN2O5S
  • Appearance: Solid
  • Color: White to off-white
  • Recommended storage: 4°C, sealed, away from moisture and light
  • Solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)
  • Solubility in DMSO: 50 mg/mL (112.37 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.