Learn More
Medchemexpress LLC Tamsulosin (hydrochloride) | 106463-17-6 | 99.8% | 25 MG
SDP
Catalog No. 5000450747
Encompass_Preferred
Tamsulosin hydrochloride is an orally active antagonist of the α1-adrenergic receptor. It induces apoptosis and is used for research related to prostatic hyperplasia. It also attenuates abdominal aortic aneurysm growth and inhibits inflammation in animal models.
  • Orally active α1-adrenergic receptor antagonist
  • Induces apoptosis
  • Used for research of prostatic hyperplasia
  • Attenuates abdominal aortic aneurysm growth
  • Inhibits inflammation in animal models
  • Stable under recommended storage conditions
  • White to off-white solid appearance
Catalog No. 50-004-50747 Supplier Medchemexpress LLC Supplier No. 25MGHYB0661A
Tamsulosin hydrochloride is an orally active antagonist of the α1-adrenergic receptor. It induces apoptosis and is used for research related to prostatic hyperplasia. It also attenuates abdominal aortic aneurysm growth and inhibits inflammation in animal models.
  • Orally active α1-adrenergic receptor antagonist
  • Induces apoptosis
  • Used for research of prostatic hyperplasia
  • Attenuates abdominal aortic aneurysm growth
  • Inhibits inflammation in animal models
  • Stable under recommended storage conditions
  • White to off-white solid appearance