Learn More
TARGETMOL CHEMICALS INC TAMSULOSIN HYDROCHLORIDE 10MG
SDP
Catalog No. 502577460
Encompass_Preferred
Also available in 25mg and bulk. Please contact Fisher for quotes. Tamsulosin hydrochloride (Flomax hydrochloride) is the hydrochloride salt of tamsulosin a sulfonamide derivative with α1 adrenergic antagonist activity. purity: 99%
Catalog No. 50-257-7460 Supplier TARGETMOL CHEMICALS INC Supplier No. T021010MG
Also available in 25mg and bulk. Please contact Fisher for quotes. Tamsulosin hydrochloride (Flomax hydrochloride) is the hydrochloride salt of tamsulosin a sulfonamide derivative with α1 adrenergic antagonist activity. purity: 99%