Learn More
Selleck Chemical LLC Tamsulosin hydrochloride 25mg 106463-17-6 Flomax hydrochloride
SDP
Supplier:  Selleck Chemical LLC S143525MG
Encompass_Preferred
Tamsulosin (Flomax) hydrochloride is an antagonist of alpha1A adrenoceptors in the prostate. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8360