Learn More
Cayman Chemical TamsulosInd4hydroCLOrID 2.5mg
SDP
Catalog No. 502939379
Encompass_Preferred
An internal standard for the quantification of tamsulosin by GC- or LC-MS
Catalog No. 50-293-9379 Supplier Cayman Chemical Supplier No. 284862.5 MG
An internal standard for the quantification of tamsulosin by GC- or LC-MS