Learn More
Uline Rubber Bands Tan, 7x1/8"""", 200/pack, 2 packs/case
Catalog No. NC1534171
Encompass
Latex rubber bands tan 7 x 1/8"""", high tensile strength, 700% elongation, 55% latex
Catalog No. NC1534171 Supplier Uline Supplier No. S11490T
Latex rubber bands tan 7 x 1/8"""", high tensile strength, 700% elongation, 55% latex