Learn More
Aqua Solutions Total Acid Number (TAN) Standard, 0.1 mg KOH/gm, Size 500ml
SDP
Catalog No. NC2238555
Encompass_Preferred
The Total Acid Number (TAN) Standard 0.1 mg KOH/gm is a 500mL standard solution used for calibration and performance verification of TAN testing. It contains 2.5 milligrams of potassium hydroxide per gram of sample dissolved in xylene. This standard allows for accurate TAN measurement and monitoring of oil degradation. The TAN Standard has a 12 month shelf life and is manufactured in an ISO 9001 certified facility. It comes with a Certificate of Analysis for traceability. This product enables proper calibration for TAN testing.
This item is not returnable. View return policy
Catalog No. NC2238555 Supplier Aqua Solutions Supplier No. 9141500ML
The Total Acid Number (TAN) Standard 0.1 mg KOH/gm is a 500mL standard solution used for calibration and performance verification of TAN testing. It contains 2.5 milligrams of potassium hydroxide per gram of sample dissolved in xylene. This standard allows for accurate TAN measurement and monitoring of oil degradation. The TAN Standard has a 12 month shelf life and is manufactured in an ISO 9001 certified facility. It comes with a Certificate of Analysis for traceability. This product enables proper calibration for TAN testing.