Learn More
Aqua Solutions TAN STD. 2.5MGKOH/G 500ML
SDP
Catalog No. NC0443667
Encompass_Preferred
TOTAL ACID NUMBER (TAN) STANDARD 2.5 mg KOH/gm, PREPARED TO ASTM D664-07 (500ML)
Catalog No. NC0443667 Supplier Aqua Solutions Supplier No. 9140500ML
TOTAL ACID NUMBER (TAN) STANDARD 2.5 mg KOH/gm, PREPARED TO ASTM D664-07 (500ML)