Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Clc Electrician's Tool Pouch, Top Grain Leather, 8-Pocket, Tan, 1 ea

Catalog No. 502769383
Encompass_Preferred

This top grain leather electrician's tool pouch provides organized storage for essential equipment. It features one large main pocket paired with six smaller pockets and sleeves to secure tools, along with specialized attachments for enhanced utility.

  • Material: Top grain leather
  • Pockets: 8 total pockets
  • Dimensions: 10-1/4 in. H x 6-1/2 in. W x 4-3/4 in. D
  • Belt compatibility: Fits belts up to 2-3/4 in. wide
  • Storage capacity: Includes hammer loop, tape measure holder (fits up to 30 ft. tapes), electrical tape chain, and metal tool clip
  • Color: Tan

Catalog No. 50-276-9383 Supplier Grainger Supplier No. 13R626
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This top grain leather electrician's tool pouch provides organized storage for essential equipment. It features one large main pocket paired with six smaller pockets and sleeves to secure tools, along with specialized attachments for enhanced utility.

  • Material: Top grain leather
  • Pockets: 8 total pockets
  • Dimensions: 10-1/4 in. H x 6-1/2 in. W x 4-3/4 in. D
  • Belt compatibility: Fits belts up to 2-3/4 in. wide
  • Storage capacity: Includes hammer loop, tape measure holder (fits up to 30 ft. tapes), electrical tape chain, and metal tool clip
  • Color: Tan

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.