Learn More
CAS DATALOGGERS TandD RTR-500BW WiFi and Ethernet Base Station for 500 Series Data Loggers
Catalog No. NC3847149
Encompass_Preferred
"The RTR-500BW is a wireless data collector (base unit) with built-in wireless/wired LAN communication capabilities. It collects data from nearby data loggers via wireless radio communication and automatically sends data via LAN to the free cloud storage service “T&D WebStorage Service” using HTTPS.It also has a warning monitoring function that sends a warning e-mail via WebStorage Service when a set limit has been exceeded. As long as there is a LAN environment, the RTR-500BW will do the data management work for you.The RTR-500BW can be combined with any of our RTR-500B/RTR-500 series wireless data loggers. Up to 50 loggers can be registered for simultaneous use."
Catalog No. NC3847149 Supplier CAS DATALOGGERS Supplier No. RTR500BW
"The RTR-500BW is a wireless data collector (base unit) with built-in wireless/wired LAN communication capabilities. It collects data from nearby data loggers via wireless radio communication and automatically sends data via LAN to the free cloud storage service “T&D WebStorage Service” using HTTPS.It also has a warning monitoring function that sends a warning e-mail via WebStorage Service when a set limit has been exceeded. As long as there is a LAN environment, the RTR-500BW will do the data management work for you.The RTR-500BW can be combined with any of our RTR-500B/RTR-500 series wireless data loggers. Up to 50 loggers can be registered for simultaneous use."