Learn More
CAS DATALOGGERS TandD RTR-501BL Wireless Temperature Data Logger with Internal Sensor & 4 Year Battery Life (Requires Base Station)
Catalog No. NC3847139
Encompass_Preferred
"The RTR501BL is a compact rugged temperature data logger with an internal temperature sensor and slow response to changes in temperature. It is capable of reading temperatures in environments of -40C to 80C.It also features a waterproof rating of IP67 (immersion proof) and an extended battery life of up to 4 years.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR-501B is comparatively slow to follow changes in temperature making it perfect for temperature control of products stored in the refrigerator, warehouses, walk in coolers, or in cooled environments.REQUIRES A BASE STATION (NOT INCLUDED)."
Catalog No. NC3847139 Supplier CAS DATALOGGERS Supplier No. RTR501BL
"The RTR501BL is a compact rugged temperature data logger with an internal temperature sensor and slow response to changes in temperature. It is capable of reading temperatures in environments of -40C to 80C.It also features a waterproof rating of IP67 (immersion proof) and an extended battery life of up to 4 years.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR-501B is comparatively slow to follow changes in temperature making it perfect for temperature control of products stored in the refrigerator, warehouses, walk in coolers, or in cooled environments.REQUIRES A BASE STATION (NOT INCLUDED)."