Learn More
CAS DATALOGGERS TandD RTR-503BL Wireless Temperature and Humidity Data Logger with 4 Year Battery Life (Requires Base Station)
Catalog No. NC3847142
Encompass_Preferred
"The RTR503BL is a temperature and humidity data logger with an all-in-one external sensor that measures temperatures from 0-55C and humidity from 10-95% RH.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR503BL with its wireless radio communication capabilities is perfect for applications where temperature and humidity at various measurement points are managed at once, such as in temperature-controlled storage facilities and warehouses.REQUIRES A BASE STATION (NOT INCLUDED)."
Catalog No. NC3847142 Supplier CAS DATALOGGERS Supplier No. RTR503BL
"The RTR503BL is a temperature and humidity data logger with an all-in-one external sensor that measures temperatures from 0-55C and humidity from 10-95% RH.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR503BL with its wireless radio communication capabilities is perfect for applications where temperature and humidity at various measurement points are managed at once, such as in temperature-controlled storage facilities and warehouses.REQUIRES A BASE STATION (NOT INCLUDED)."