Learn More
CAS DATALOGGERS TandD RTR-505BL-TC Wireless Temperature Data Logger with Insulated Thermocouple Probe & 4 Year Battery Life (Requires Base Station)
Catalog No. NC3847147
Encompass_Preferred
"The RTR505BL-TC is a highly versatile thermocouple data logger that can measure a wide range from -200 to 1760C. This device comes with a sealed type K thermocouple that measures from -200C to 200C.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR505BL-TC with its wireless radio communication capabilities is perfect for applications where strict temperature control is required, such as for pharmaceuticals and laboratories. It is also well equipped to handle cryogenic temperature measurements as it can measure down to -200C.REQUIRES A BASE STATION (NOT INCLUDED)."
Catalog No. NC3847147 Supplier CAS DATALOGGERS Supplier No. RTR505BLTCKTYPE
"The RTR505BL-TC is a highly versatile thermocouple data logger that can measure a wide range from -200 to 1760C. This device comes with a sealed type K thermocouple that measures from -200C to 200C.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR505BL-TC with its wireless radio communication capabilities is perfect for applications where strict temperature control is required, such as for pharmaceuticals and laboratories. It is also well equipped to handle cryogenic temperature measurements as it can measure down to -200C.REQUIRES A BASE STATION (NOT INCLUDED)."