Learn More
CAS DATALOGGERS TandD RTR-507BL Wireless High Accuracy Temperature and Humidity Data Logger with 4 Year Battery Life (Requires Base Station)
Catalog No. NC3847144
Encompass_Preferred
"The RTR507BL is a high accuracy/prevision temperature and humidity data logger with an all-in-one external sensor that measures temperatures from -25C to 70C and humidity from 0-99% RH.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR507BL with its wireless radio communication capabilities is perfect for applications where strict temperature and humidity control is required, such as for pharmaceuticals and laboratories."
Catalog No. NC3847144 Supplier CAS DATALOGGERS Supplier No. RTR507BL
"The RTR507BL is a high accuracy/prevision temperature and humidity data logger with an all-in-one external sensor that measures temperatures from -25C to 70C and humidity from 0-99% RH.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR507BL with its wireless radio communication capabilities is perfect for applications where strict temperature and humidity control is required, such as for pharmaceuticals and laboratories."