Learn More
CAS DATALOGGERS TandD RTR-576 Wireless Carbon Dioxide, Temperature, and Humidity Data Logger (Requires Base Station)
Catalog No. NC3847148
Encompass_Preferred
"The RTR-576 is a three-channel data logger designed to measure and record CO2 concentration, temperature and humidity.External devices such as buzzers and lights can also be connected to the contact output terminal to provide instant on-site notification, signaling when ventilation may be needed.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR505BL-TC with its wireless radio communication capabilities is perfect for applications where CO2 has to be managed and monitored.REQUIRES A BASE STATION (NOT INCLUDED)."
Catalog No. NC3847148 Supplier CAS DATALOGGERS Supplier No. RTR576
"The RTR-576 is a three-channel data logger designed to measure and record CO2 concentration, temperature and humidity.External devices such as buzzers and lights can also be connected to the contact output terminal to provide instant on-site notification, signaling when ventilation may be needed.Data can be transmitted from a logger to a base unit and then automatically uploaded to the free cloud storage software or saved to a specified location. Data can also be checked on your PC screen or on-site with one of our compatible data collectors.The RTR505BL-TC with its wireless radio communication capabilities is perfect for applications where CO2 has to be managed and monitored.REQUIRES A BASE STATION (NOT INCLUDED)."