Learn More
CAS DATALOGGERS TandD TR72A2 WiFi/Bluetooth Temperature and Humidity Data Logger
Catalog No. NC3847112
Encompass_Preferred
"The TR72A2 allows you to measure and record temperature from 0 to 55°C and humidity from 10 to 95%RH. The LCD screen allows the simultaneous display of either the current values of two channels, the highest and lowest values of channel 1, or the highest and lowest values of channel 2. The ALM icon on the LCD alerts the user of the occurrence of an alarm.Use the wireless feature to auto-upload data to the free cloud software for anytime, anywhere access, receive warning notifications, and generate reports. You can configure device settings, generate reports, download data, and view the uploaded data using Bluetooth or USB.This kit includes the device, a mounting bracket, USB cable, and USB power outlet."
Catalog No. NC3847112 Supplier CAS DATALOGGERS Supplier No. TR72A2KIT
"The TR72A2 allows you to measure and record temperature from 0 to 55°C and humidity from 10 to 95%RH. The LCD screen allows the simultaneous display of either the current values of two channels, the highest and lowest values of channel 1, or the highest and lowest values of channel 2. The ALM icon on the LCD alerts the user of the occurrence of an alarm.Use the wireless feature to auto-upload data to the free cloud software for anytime, anywhere access, receive warning notifications, and generate reports. You can configure device settings, generate reports, download data, and view the uploaded data using Bluetooth or USB.This kit includes the device, a mounting bracket, USB cable, and USB power outlet."