Learn More
CAS DATALOGGERS TandD TR72NW Ethernet Temperature and Humidity Data Logger
Catalog No. NC3847121
Encompass_Preferred
"The TR72NW allows you to measure and record temperature from 0 to 55°C and humidity from 10 to 95%RH. The LCD screen allows the simultaneous display of either the current values of two channels, the highest and lowest values of channel 1, or the highest and lowest values of channel 2. The ALM icon on the LCD alerts the user of the occurrence of an alarm.Use the Ethernet feature to auto-upload data to the free cloud software for anytime, anywhere access, receive warning notifications, and generate reports. You can configure device settings, generate reports, download data, and view the uploaded data using a USB cord.This kit includes the device, a mounting bracket, USB cable, and USB power outlet."
Catalog No. NC3847121 Supplier CAS DATALOGGERS Supplier No. TR72NWKIT
"The TR72NW allows you to measure and record temperature from 0 to 55°C and humidity from 10 to 95%RH. The LCD screen allows the simultaneous display of either the current values of two channels, the highest and lowest values of channel 1, or the highest and lowest values of channel 2. The ALM icon on the LCD alerts the user of the occurrence of an alarm.Use the Ethernet feature to auto-upload data to the free cloud software for anytime, anywhere access, receive warning notifications, and generate reports. You can configure device settings, generate reports, download data, and view the uploaded data using a USB cord.This kit includes the device, a mounting bracket, USB cable, and USB power outlet."