Learn More
CAS DATALOGGERS TandD TR75NW Ethernet Temperature Data Logger with 2 Sealed Thermocouple Probes
Catalog No. NC3847133
Encompass_Preferred
"The TR75NW is a temperature data logger that can connect up to 2 thermocouple (K, J, T, E, S, R) sensors. It is capable of measuring temperatures ranging from -199 to 1760°C and can be used in very high temperatures such as in a plant furnace or in deep freezers where temperatures reach very low levels.Use the Ethernet feature to auto-upload data to the free cloud software for anytime, anywhere access, receive warning notifications, and generate reports. You can configure device settings, generate reports, download data, and view the uploaded data using a USB cord.This kit includes the device, 2 3ft sealed thermocouple probes for measuring (-200C to 100C), 2 thermal buffers, a mounting bracket, USB cable, and USB power outlet."
Catalog No. NC3847133 Supplier CAS DATALOGGERS Supplier No. TR75NWKIT2
"The TR75NW is a temperature data logger that can connect up to 2 thermocouple (K, J, T, E, S, R) sensors. It is capable of measuring temperatures ranging from -199 to 1760°C and can be used in very high temperatures such as in a plant furnace or in deep freezers where temperatures reach very low levels.Use the Ethernet feature to auto-upload data to the free cloud software for anytime, anywhere access, receive warning notifications, and generate reports. You can configure device settings, generate reports, download data, and view the uploaded data using a USB cord.This kit includes the device, 2 3ft sealed thermocouple probes for measuring (-200C to 100C), 2 thermal buffers, a mounting bracket, USB cable, and USB power outlet."