Learn More
WELCH MATERIALS INC TandexTM G25M, 100g/bottle
Catalog No. 502093189
Encompass_Preferred
Name: Tandex G25M, specification: 100g, property: gel giltration media(highly cross-linked), bead structure: dextran, mean particle size(dry): 50-150 μm, exclusion range(globulin, Mr): 1,000-5,000, Sterilization: 121℃ in water for 20min, pH stability: 2-12(long term), 2-14(short term), flow rate: 150-250 cm/h(column height 10cm, dia: 5cm, 0.1Mpa, 25°C), storage condition: 20% ethanol, 4-30°C. Similar Products: Sephadex G-25 Medium
Catalog No. 50-209-3189 Supplier WELCH MATERIALS INC Supplier No. W1062
Name: Tandex G25M, specification: 100g, property: gel giltration media(highly cross-linked), bead structure: dextran, mean particle size(dry): 50-150 μm, exclusion range(globulin, Mr): 1,000-5,000, Sterilization: 121℃ in water for 20min, pH stability: 2-12(long term), 2-14(short term), flow rate: 150-250 cm/h(column height 10cm, dia: 5cm, 0.1Mpa, 25°C), storage condition: 20% ethanol, 4-30°C. Similar Products: Sephadex G-25 Medium