Learn More
Biolegend TANDEM STABILIZER HELPS TO RED
Catalog No. NC2184113
Encompass_Preferred
Tandem Stabilizer helps to reduce tandem dye de-coupling when added as a supplement to buffers commonly used to store stained, fixed cells, as well as those used for fixation and permeabilization procedures.
Catalog No. NC2184113 Supplier Biolegend Supplier No. 421802
Tandem Stabilizer helps to reduce tandem dye de-coupling when added as a supplement to buffers commonly used to store stained, fixed cells, as well as those used for fixation and permeabilization procedures.