Learn More
TARGETMOL CHEMICALS INC TANDOSPIRONE CITRATE 25MG
SDP
Catalog No. 502579192
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tandospirone citrate (SM-3997 citrate) is a potent and selective 5-HT1A receptor partial agonist (Ki 27 nM)purity: 99%
Catalog No. 50-257-9192 Supplier TARGETMOL CHEMICALS INC Supplier No. T741225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tandospirone citrate (SM-3997 citrate) is a potent and selective 5-HT1A receptor partial agonist (Ki 27 nM)purity: 99%