Learn More
Abcam Tandospirone hydrochloride (Metanopirone), 5-HT1A partial agonist, 10MG
Catalog No. 502704936
Encompass_Preferred
MW 419.9 Da, Purity >99%. Potent, selective 5-HT1A partial agonist (Ki = 27 nM). Decreases the firing rate of 5-HT neurons of the dorsal raphe. Shows anxiolytic and antidepressant effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4936 Supplier Abcam Supplier No. AB14613610MG
Add to Cart
Add to Cart
MW 419.9 Da, Purity >99%. Potent, selective 5-HT1A partial agonist (Ki = 27 nM). Decreases the firing rate of 5-HT neurons of the dorsal raphe. Shows anxiolytic and antidepressant effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement