Learn More
Medchemexpress LLC TANDSOPIRONE 50.0MG
SDP
Catalog No. NC3806144
Encompass_Preferred
Tandospirone /SM-3997/ is a potent and selective 5-HT1A receptor partial agonist with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.For research use only.
Catalog No. NC3806144 Supplier Medchemexpress LLC Supplier No. HY14558
Tandospirone /SM-3997/ is a potent and selective 5-HT1A receptor partial agonist with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.For research use only.