Learn More
TARGETMOL CHEMICALS INC Tandutinib 200MG
SDP
Catalog No. 502876078
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tandutinib (CT53518) (MLN518, CT53518), an effective FLT3 antagonist (IC50 0.22 uM), can also inhibit c-Kit and PDGFR, 15-20 fold higher potency for FLT3 versus CSF-1R and >100-fold selectivity for the same target versus FGFR, EGFR, and KDR. Purity 99.74%
Catalog No. 50-287-6078 Supplier TARGETMOL CHEMICALS INC Supplier No. T1667200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tandutinib (CT53518) (MLN518, CT53518), an effective FLT3 antagonist (IC50 0.22 uM), can also inhibit c-Kit and PDGFR, 15-20 fold higher potency for FLT3 versus CSF-1R and >100-fold selectivity for the same target versus FGFR, EGFR, and KDR. Purity 99.74%