Learn More
Medchemexpress LLC HY-10202A 100mg Medchemexpress, Tandutinib (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10202A100MG
Encompass_Preferred
Medchemexpress, HY-10202A 100mg Tandutinib (HCl) CAS: Tandutinib hydrochloride (MLN518 hydrochloride) is a potent and selective inhibitor of the FLT3 with an IC50 of 0.22 μM, and also inhibits c-Kit and PDGFR with IC50s of 0.17 μM and 0.20 μM, respectively. Tandutinib hydrochloride can be used for acute myelogenous leukemia (AML). Tandutinib hydrochloride has the ability to cross the blood-brain barrier[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7957