Learn More
Medchemexpress LLC Tandutinib hydrochloride | 2438900-70-8 | 99.3% | 599.16 g/mol | C31H43ClN6O4 | 1 ML
SDP
Catalog No. 5000247372
Encompass_Preferred
Tandutinib hydrochloride is a small-molecule tyrosine kinase inhibitor selective for FLT3, with additional activity against c-Kit and PDGFR. It is used as a research reagent in biochemical and cell-based assays to probe signaling pathways and assess antiproliferative effects in oncology studies. The compound has defined physicochemical properties and is suitable for preparing stock solutions for in vitro workflows.
  • Potent FLT3 inhibition (IC50 ≈ 0.22 μM).
  • Also inhibits c-Kit and PDGFR, supporting broader kinase profiling.
  • High purity suitable for research applications (>99% by HPLC).
  • Suitable for preparing accurate stock solutions for assays (e.g., 10 mM).
  • Provided with datasheet information for safe handling and solubility guidance.
Catalog No. 50-002-47372 Supplier Medchemexpress LLC Supplier No. HY10202A10MM 1ML
Tandutinib hydrochloride is a small-molecule tyrosine kinase inhibitor selective for FLT3, with additional activity against c-Kit and PDGFR. It is used as a research reagent in biochemical and cell-based assays to probe signaling pathways and assess antiproliferative effects in oncology studies. The compound has defined physicochemical properties and is suitable for preparing stock solutions for in vitro workflows.
  • Potent FLT3 inhibition (IC50 ≈ 0.22 μM).
  • Also inhibits c-Kit and PDGFR, supporting broader kinase profiling.
  • High purity suitable for research applications (>99% by HPLC).
  • Suitable for preparing accurate stock solutions for assays (e.g., 10 mM).
  • Provided with datasheet information for safe handling and solubility guidance.