Learn More
Apexbio Technology LLC Tandutinib (MLN518) 387867-13-2 200mg
SDP
Catalog No. 5000092184
Encompass_Preferred
Tandutinib (MLN518 CAS 387867-13-2) is a selective small molecule inhibitor primarily targeting FLT3 with additional activity against PDGFR and c-Kit In biochemical kinase assays Tandutinib exhibits IC50 values of 0 22 M for FLT3 0 20 M for PDGFR and 0 17 M for c-Kit It effectively inhibits tyrosine phosphorylation of both wild-type FLT3 and FLT3 mutants with IC50 values in the 30 100 nM range In cellular models Tandutinib demonstrates anti-proliferative effects in FLT3-ITD-positive cell lines (Molm-13 Molm-14) at low nanomolar concentrations whereas FLT3-ITD-negative cells are substantially less sensitive This compound is utilized in research investigating FLT3-mediated signaling and acute myelogenous leukemia (AML) pathogenesis
Catalog No. 50-000-92184 Supplier Apexbio Technology LLC Supplier No. B1526200
Tandutinib (MLN518 CAS 387867-13-2) is a selective small molecule inhibitor primarily targeting FLT3 with additional activity against PDGFR and c-Kit In biochemical kinase assays Tandutinib exhibits IC50 values of 0 22 M for FLT3 0 20 M for PDGFR and 0 17 M for c-Kit It effectively inhibits tyrosine phosphorylation of both wild-type FLT3 and FLT3 mutants with IC50 values in the 30 100 nM range In cellular models Tandutinib demonstrates anti-proliferative effects in FLT3-ITD-positive cell lines (Molm-13 Molm-14) at low nanomolar concentrations whereas FLT3-ITD-negative cells are substantially less sensitive This compound is utilized in research investigating FLT3-mediated signaling and acute myelogenous leukemia (AML) pathogenesis