Learn More
Selleck Chemical LLC Tandutinib (MLN518) 25mg 387867-13-2 CT 53518, NSC726292, MLN518
SDP
Catalog No. 508267
Encompass_Preferred
Tandutinib (MLN518, CT53518, NSC726292) is a potent FLT3 antagonist with IC50 of 0.22 ?M, also inhibits PDGFR and c-Kit, 15 to 20-fold higher potency for FLT3 versus CSF-1R and >100-fold selectivity for the same target versus FGFR, EGFR and KDR. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-826-7 Supplier Selleck Chemical LLC Supplier No. S104325MG
Tandutinib (MLN518, CT53518, NSC726292) is a potent FLT3 antagonist with IC50 of 0.22 ?M, also inhibits PDGFR and c-Kit, 15 to 20-fold higher potency for FLT3 versus CSF-1R and >100-fold selectivity for the same target versus FGFR, EGFR and KDR. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.