Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tanespimycin (17-AAG) | 75747-14-7 | MFCD04973892 | 99.0% | 585.69 g/mol | C31H43N3O8 | 5 MG
SDP

Catalog No. 5000247062
Encompass_Preferred

Tanespimycin (17-AAG) is a small-molecule inhibitor of heat shock protein 90 (HSP90) used for in vitro and in vivo research. It is supplied as a high-purity research chemical with manufacturer guidance for storage and formulation.

  • Potent HSP90 inhibitor with low-nanomolar activity.
  • Provided as a high-purity research compound suitable for experimental work.
  • Available in small research quantities, including 5 mg vials.
  • Storage: store at 4°C; in solvent, store at -80°C for long-term stability.
  • Solubility: soluble in DMSO; manufacturer provides in vivo formulation guidance for PEG300, SBE-β-CD, and Cremophor EL.

Catalog No. 50-002-47062 Supplier Medchemexpress LLC Supplier No. HY102115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tanespimycin (17-AAG) is a small-molecule inhibitor of heat shock protein 90 (HSP90) used for in vitro and in vivo research. It is supplied as a high-purity research chemical with manufacturer guidance for storage and formulation.

  • Potent HSP90 inhibitor with low-nanomolar activity.
  • Provided as a high-purity research compound suitable for experimental work.
  • Available in small research quantities, including 5 mg vials.
  • Storage: store at 4°C; in solvent, store at -80°C for long-term stability.
  • Solubility: soluble in DMSO; manufacturer provides in vivo formulation guidance for PEG300, SBE-β-CD, and Cremophor EL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.