Learn More
Abcam Tanespimycin (17-AAG), HSP90 inhibitor, 5MG
Catalog No. 502706619
Encompass_Preferred
MW 585.7 Da, Purity >98%. Potent, selective HSP90 inhibitor (IC₅₀ = 5 nM). Anticancer activity, including cytostatic, proapoptotic and antiangiogenic properties in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6619 Supplier Abcam Supplier No. AB1414335MG
MW 585.7 Da, Purity >98%. Potent, selective HSP90 inhibitor (IC₅₀ = 5 nM). Anticancer activity, including cytostatic, proapoptotic and antiangiogenic properties in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement