Learn More
Medchemexpress LLC HY-10211 25mg , Tanespimycin CAS:75747-14-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1021125MG
Encompass_Preferred
HY-10211 25mg Tanespimycin CAS: 75747-14-7 Tanespimycin (17-AAG) is a potent HSP90 inhibitor with an IC50 of 5 nM, having a 100-fold higher binding affinity for tumour cell derived HSP90 than normal cell derived HSP90[5]. Tanespimycin depletes cellular STK38/NDR1 and reduces STK38 kinase activity. Tanespimycin also downregulates the stk38 gene expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0431