Learn More
Selleck Chemical LLC Tanespimycin (17-AAG) S1141-5mg
SDP
Catalog No. 503108225
Encompass_Preferred
Tanespimycin (17-AAG CP127374 NSC-330507 KOS 953) is a potent HSP90 inhibitor with IC50 of 5 nM in a cell-free assay having a 100-fold higher binding affinity for HSP90 derived from tumour cells than HSP90 from normal cells Tanespimycin (17-AAG) induces apoptosis necrosis autophagy and mitophagy Phase 3
Catalog No. 50-310-8225 Supplier Selleck Chemical LLC Supplier No. S11415MG
Tanespimycin (17-AAG CP127374 NSC-330507 KOS 953) is a potent HSP90 inhibitor with IC50 of 5 nM in a cell-free assay having a 100-fold higher binding affinity for HSP90 derived from tumour cells than HSP90 from normal cells Tanespimycin (17-AAG) induces apoptosis necrosis autophagy and mitophagy Phase 3