Learn More
REPLIGEN CORPORATION SIUS PD 0.02m2 (LP) PrS 30kD Cassette
SDP
Supplier:  REPLIGEN CORPORATION PP030LP2L
Encompass
SIUS PD cassette with ProStream 30kD L-screen 0.02m2 holder-L.
Catalog No. NC1563163