Learn More
Medchemexpress LLC HY-N0133 10mg Medchemexpress, Tangeretin CAS:481-53-8 Purity:>98%
SDP
Catalog No. 501933415
Encompass_Preferred
Medchemexpress, HY-N0133 10mg Tangeretin CAS:481-53-8 Tangeretin (Tangeritin), a flavonoid from citrus fruit peels, has been proven to play an important role in anti-inflammatory responses and neuroprotective effects in several disease models, and was also selected as a Notch-1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3415 Supplier Medchemexpress LLC Supplier No. HYN013310MG
Medchemexpress, HY-N0133 10mg Tangeretin CAS:481-53-8 Tangeretin (Tangeritin), a flavonoid from citrus fruit peels, has been proven to play an important role in anti-inflammatory responses and neuroprotective effects in several disease models, and was also selected as a Notch-1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.