Learn More
Selleck Chemical LLC Tangeretin S2363-5mg
SDP
Catalog No. 503112710
Encompass_Preferred
Tangeretin (Tangeritin) a natural polymethoxylated flavone concentrated in the peel of citrus fruits an inhibitor of Notch1 is known to have antiproliferative antiinvasive antimetastatic and antioxidant activities
Catalog No. 50-311-2710 Supplier Selleck Chemical LLC Supplier No. S23635MG
Tangeretin (Tangeritin) a natural polymethoxylated flavone concentrated in the peel of citrus fruits an inhibitor of Notch1 is known to have antiproliferative antiinvasive antimetastatic and antioxidant activities