Learn More
Apexbio Technology LLC Tangeretin 481-53-8 20mg
SDP
Catalog No. 5000094444
Encompass_Preferred
Tangeretin (CAS 481-53-8) is a polymethoxyflavone isolated from the peel of Citrus nobilis This compound has been shown to modulate various cellular signaling pathways including inhibition of phosphoinositide 3-kinase/Akt and MAPK pathways thus influencing processes such as cell proliferation differentiation and apoptosis Tangeretin has been investigated for its anti-inflammatory antioxidant and anticancer properties in preclinical models Due to its broad spectrum of biological activities Tangeretin is utilized in research exploring the molecular mechanisms underlying inflammation oxidative stress and tumorigenesis Store in airtight containers away from heat and moisture
Catalog No. 50-000-94444 Supplier Apexbio Technology LLC Supplier No. N207720
Tangeretin (CAS 481-53-8) is a polymethoxyflavone isolated from the peel of Citrus nobilis This compound has been shown to modulate various cellular signaling pathways including inhibition of phosphoinositide 3-kinase/Akt and MAPK pathways thus influencing processes such as cell proliferation differentiation and apoptosis Tangeretin has been investigated for its anti-inflammatory antioxidant and anticancer properties in preclinical models Due to its broad spectrum of biological activities Tangeretin is utilized in research exploring the molecular mechanisms underlying inflammation oxidative stress and tumorigenesis Store in airtight containers away from heat and moisture