Learn More
Cayman Chemical Tangeretin Neurochemicals
SDP
Catalog No. 502888220
Encompass_Preferred
A polymethoxylated flavone isolated from citrus peels; inhibits signaling in cancer cells, reducing ERK signaling in T47D breast cancer cells (IC50 ~3 μM)
Catalog No. 50-288-8220 Supplier Cayman Chemical Supplier No. 100099115 MG
A polymethoxylated flavone isolated from citrus peels; inhibits signaling in cancer cells, reducing ERK signaling in T47D breast cancer cells (IC50 ~3 μM)