Learn More
Cayman Chemical Tangeretin, Purity: ≥98%, CAS: 481-53-8, 100 mg
SDP
Catalog No. 502890174
Encompass_Preferred
A polymethoxylated flavone isolated from citrus peels; inhibits signaling in cancer cells, reducing ERK signaling in T47D breast cancer cells (IC50 ~3 μM)
Catalog No. 50-289-0174 Supplier Cayman Chemical Supplier No. 10009911100 MG
A polymethoxylated flavone isolated from citrus peels; inhibits signaling in cancer cells, reducing ERK signaling in T47D breast cancer cells (IC50 ~3 μM)