Learn More
Medchemexpress LLC Taniborbactam hydrochloride | 2244235-49-0 | 100.0% | 462.18 | 1 ML
SDP
Catalog No. 5000346073
Encompass_Preferred
Taniborbactam hydrochloride (VNRX-5133 hydrochloride) is a reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor. It is active against Gram-negative bacteria and intended for research use only, not sold to patients.
  • Reversible and selective pan-spectrum β-lactamase inhibitor
  • Active against Gram-negative bacteria
  • Exhibits IC50s ranging from 8-530 nM
  • Specific IC50s include 30 nM for KPC-2, 32 nM for AmpC, 42 nM for OXA-48, and 20 nM for VIM-2
Catalog No. 50-003-46073 Supplier Medchemexpress LLC Supplier No. HY109124A1MG
Taniborbactam hydrochloride (VNRX-5133 hydrochloride) is a reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor. It is active against Gram-negative bacteria and intended for research use only, not sold to patients.
  • Reversible and selective pan-spectrum β-lactamase inhibitor
  • Active against Gram-negative bacteria
  • Exhibits IC50s ranging from 8-530 nM
  • Specific IC50s include 30 nM for KPC-2, 32 nM for AmpC, 42 nM for OXA-48, and 20 nM for VIM-2