Learn More
Medchemexpress LLC Taniborbactam (hydrochloride) | 2244235-49-0 | 100.0% | 462.18 | 50 MG
SDP
Catalog No. 5000365875
Encompass_Preferred
Taniborbactam hydrochloride (VNRX-5133 hydrochloride) is a reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor with IC50s of 8-530 nM. It is active against Gram-negative bacteria and effective against various β-lactamases, including KPC-2, AmpC, OXA-48, and VIM-2. This product is for research use only.
  • Reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor.
  • Active against gram-negative bacteria.
  • Effective against KPC-2, AmpC, OXA-48, and VIM-2 β-lactamases.
  • Demonstrated efficacy in neutropenic mouse lung infection and urinary tract infection models.
  • Used in combination with cefepime or meropenem for enhanced activity against NDM-1-producing clinical isolates.
  • Available documentation includes datasheet, COA, SDS, HNMR, and RP-HPLC.
  • For research use only.
Catalog No. 50-003-65875 Supplier Medchemexpress LLC Supplier No. 50MGHY109124A
Taniborbactam hydrochloride (VNRX-5133 hydrochloride) is a reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor with IC50s of 8-530 nM. It is active against Gram-negative bacteria and effective against various β-lactamases, including KPC-2, AmpC, OXA-48, and VIM-2. This product is for research use only.
  • Reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor.
  • Active against gram-negative bacteria.
  • Effective against KPC-2, AmpC, OXA-48, and VIM-2 β-lactamases.
  • Demonstrated efficacy in neutropenic mouse lung infection and urinary tract infection models.
  • Used in combination with cefepime or meropenem for enhanced activity against NDM-1-producing clinical isolates.
  • Available documentation includes datasheet, COA, SDS, HNMR, and RP-HPLC.
  • For research use only.