Learn More
Medchemexpress LLC Taniborbactam hydrochloride | 2244235-49-0 | 99.97% | 462.18 | 10 MG
SDP
Catalog No. 5000344493
Encompass_Preferred
Taniborbactam hydrochloride (VNRX-5133 hydrochloride) is a reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor with IC50s of 8-530 nM. It has IC50s of 30 nM, 32 nM, 42 nM, and 20 nM for KPC-2, AmpC, OXA-48, and VIM-2, respectively, and is active against Gram-negative bacteria.
  • Reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor.
  • Effective against KPC-2, AmpC, OXA-48, and VIM-2.
  • Active against Gram-negative bacteria.
Catalog No. 50-003-44493 Supplier Medchemexpress LLC Supplier No. HY109124A10MM/1ML
Taniborbactam hydrochloride (VNRX-5133 hydrochloride) is a reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor with IC50s of 8-530 nM. It has IC50s of 30 nM, 32 nM, 42 nM, and 20 nM for KPC-2, AmpC, OXA-48, and VIM-2, respectively, and is active against Gram-negative bacteria.
  • Reversible and selective boronic acid-containing pan-spectrum β-lactamase inhibitor.
  • Effective against KPC-2, AmpC, OXA-48, and VIM-2.
  • Active against Gram-negative bacteria.