Learn More
Genscript Corporation Tank Adaptor
Catalog No. NC1395872
Encompass
The adaptor is for use with Novex XCell I, II & SureLock gel tanks when running ExpressPlusᵀᴹ or SurePAGEᵀᴹ gels (8x10cm), see gel manual for detail.
Catalog No. NC1395872 Supplier Genscript Corporation Supplier No. L00671
The adaptor is for use with Novex XCell I, II & SureLock gel tanks when running ExpressPlusᵀᴹ or SurePAGEᵀᴹ gels (8x10cm), see gel manual for detail.