Learn More
Grainger Approved Vendor Tank Drain Kit, 1/4 in. Thread, For Air Compressors, 1 ea
Catalog No. 502787303
Encompass_Preferred
This tank drain kit is designed for use with portable and electric air compressors to facilitate efficient drainage. It is engineered for compatibility with specific air compressor models and ensures reliable performance for maintenance.
  • Compatible with 5P820, 6AYW7, 6GD06, and 6P769 air compressor models
  • Fits 1/4 in. or 3/8 in. NPT tank ports
  • Features a 1/4 in. thread size
Catalog No. 50-278-7303 Supplier Grainger Supplier No. 196XR6
This tank drain kit is designed for use with portable and electric air compressors to facilitate efficient drainage. It is engineered for compatibility with specific air compressor models and ensures reliable performance for maintenance.
  • Compatible with 5P820, 6AYW7, 6GD06, and 6P769 air compressor models
  • Fits 1/4 in. or 3/8 in. NPT tank ports
  • Features a 1/4 in. thread size