Learn More
Corning™ Tank Liners
Catalog No. MT9130060 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
560 L
Catalog No. Capacity (Metric)
MT9130060 560 L
1 options
Catalog No. MT9130060 Supplier Corning™ Supplier No. 9130060
Add to Cart
Add to Cart
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
  • Made from low-density polyethylene (LDPE)
  • Eliminate costs associated with cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate risks associated with cross contamination
  • Help extend life of reuseable tanks, resulting in reduced capital expenditures
  • Gamma irradiated, individually packaged
  • Choice of gusseted and non-gusseted available
  • Available in many sizes
TRUSTED_SUSTAINABILITY
Specifications
Material LDPE
Product Type Non-Gusseted Tank Liner
Capacity (Metric) 560 L
WARNING: Reproductive Harm - www.P65Warnings.ca.gov