Learn More
Sigma Aldrich Fine Chemicals Biosciences Tankyrase 2 849 1166 act
Catalog No. 502907048
Encompass
TNKS2 (TRF1-interacting ankyrin-related ADP-ribose polymerase 2) also known as PARP5B tanykyrase-2 or TNKL (GenBank Accession No. NM 025235) amino acids 849-1166 (end) with N-terminal GST-tag MW62kDa expressed in Sf9 cells via a Baculovirus expression system.
Catalog No. 50-290-7048 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP042410UG
TNKS2 (TRF1-interacting ankyrin-related ADP-ribose polymerase 2) also known as PARP5B tanykyrase-2 or TNKL (GenBank Accession No. NM 025235) amino acids 849-1166 (end) with N-terminal GST-tag MW62kDa expressed in Sf9 cells via a Baculovirus expression system.