Learn More
Cayman Chemical Tannc Acd 10g
SDP
Catalog No. 502994251
Encompass_Preferred
An orally bioavailable selective inhibitor of PDGFRa PDGFRB and FLT3 (IC50s 11 3 2 and 4 nM respectively) also inhibits mutant forms of these kinases including the D842V-containing form of PDGFR and D835Y and internal tandem duplication mutations of FLT3 at nanomolar concentrations
Catalog No. 50-299-4251 Supplier Cayman Chemical Supplier No. 2142110 G
An orally bioavailable selective inhibitor of PDGFRa PDGFRB and FLT3 (IC50s 11 3 2 and 4 nM respectively) also inhibits mutant forms of these kinases including the D842V-containing form of PDGFR and D835Y and internal tandem duplication mutations of FLT3 at nanomolar concentrations