Learn More
Cayman Chemical Tannc Acd 25g
SDP
Catalog No. 502993980
Encompass_Preferred
An inhibitor of the gastric H /K ATPase and an enantiomer of esomeprazole inhibits histamine-induced acid secretions in isolated rabbit gastric glands (IC50 781 UM) to a lesser extent than esomeprazole inhibits histamine-induced stomach acid secretion in rats at 30 mg/kg
Catalog No. 50-299-3980 Supplier Cayman Chemical Supplier No. 2142125 G
An inhibitor of the gastric H /K ATPase and an enantiomer of esomeprazole inhibits histamine-induced acid secretions in isolated rabbit gastric glands (IC50 781 UM) to a lesser extent than esomeprazole inhibits histamine-induced stomach acid secretion in rats at 30 mg/kg