Learn More
ATCC Bordetella pertussis; H920
Catalog No. 502381278
Encompass
The depositor states that this strain does not produce pertactin, and that there is an inversion nucleotide 74 mutation.
Catalog No. 50-238-1278 Supplier ATCC Supplier No. BAA2705
The depositor states that this strain does not produce pertactin, and that there is an inversion nucleotide 74 mutation.