Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tannic acid | 1401-55-4 | 81.7% | 10 G
SDP

Catalog No. 5000410361
Encompass_Preferred

Tannic acid is a novel hERG channel blocker with an IC50 of 3.4 μM.

  • Target: Potassium channel
  • Research areas: Inflammation/immunology, cancer
  • Appearance: Light yellow to yellow solid
  • Purity: 81.7%
  • Molecular formula: C76H52O46
  • Molecular weight: 1701.20
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-10361 Supplier Medchemexpress LLC Supplier No. HYB213610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tannic acid is a novel hERG channel blocker with an IC50 of 3.4 μM.

  • Target: Potassium channel
  • Research areas: Inflammation/immunology, cancer
  • Appearance: Light yellow to yellow solid
  • Purity: 81.7%
  • Molecular formula: C76H52O46
  • Molecular weight: 1701.20
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.